Delmhorst® PX-20 Paper Moisture Meters
Designed for quality control technicians, the PX-20 Paper Moisture Meter offers precise moisture monitoring for cores, paperboard, and corrugated stock to help maintain the highest standards in paper production
  • Accurate moisture range of 4.3%-18% for kraft paper, with alarm set points for user-selected thresholds
  • Large, easy-to-read display with auto backlight for all lighting conditions
  • Ergonomic ABS case with E-Z grip handle and large tactile buttons for comfort and control
  • Stores up to 100 readings with stats for High, Low, Average, and Standard Deviation
  • Integral contact pins penetrate up to 5/16” with optional external electrode compatibility
The PX-20 Paper Moisture Meter combines durability and accuracy in a rugged, ergonomic design tailored to the demands of the paper industry. Featuring an intuitive dashboard-like display, customizable alarms, and internal calibration checks, the PX-20 ensures reliable, accurate readings across applications. Whether you're testing cores, paperboard, or corrugated stock, the PX-20 provides the functionality and ease of use essential for professional quality control, making it a trusted tool in any paper production environment.
$490.00 - $501.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrode Type Measurement Type Description
Availability Pricing
Delmhorst PX-20 Paper Moisture Meter with 324CAS-0112 Case EW-59800-87 Mfr # PX-20/P01
COMPARE
None included Paper (4.3-18%) Paper Moisture Meter with 324CAS-0112 Case
$501.00USD / Each
Delmhorst PX-20 Paper Moisture Meter with 324CAS-0111 Case EW-59800-88 Mfr # PX-20W/CS
COMPARE
None included Paper (4.3-18%) Paper Moisture Meter with 324CAS-0111 Case
$490.00USD / Each