Diba Click-N-Seal® Fittings with Micro 6-40 UNF(M) Thread for 1/16" OD Tubing
Hand tighten fittings without tools to create a secure seal
  • When the cap clicks, you know the seal is tight
These unique self-torquing fittings create a secure seal without the use of any tools. As the fitting is tightened, the resistance of the cap increases until the correct torque is reached. Fittings are made of PEEK™ and are compact in design and provide excellent chemical resistance.
The fitting can spin freely, eliminating tubing from twisting. A recess in the fitting houses the ferrule allowing maximum thread engagement with the port. Type S ferrule system uses a two-piece ferrule that fits permanently to the outside of the tubing. Two-part ferrule is made up of an inner PEEK or ETFE sealing surface surrounded by a stainless steel lock ring with only the inner portion in the fluid path.
$129.25 - $147.75USD / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Color
Availability Pricing
Diba Click-N-Seal™ Micro Tubing End Fitting, 1/16" OD tubing, 6-40 UNF(M), black PEEK™; 10/pk EW-21942-04 Mfr # 008NF16-KT9B
COMPARE
Black
$147.75USD / Pkg of 10
Diba Click-N-Seal™ Micro Tubing End Fitting, 1/16" OD tubing, 6-40 UNF(M), natural PEEK™; 10/pk EW-21942-05 Mfr # 008NF16-KT9L
COMPARE
Natural
$129.25USD / Pkg of 10
Diba Click-N-Seal™ Micro Tubing End Fitting, 1/16" OD tubing, 6-40 UNF(M), red PEEK™; 10/pk EW-21942-06 Mfr # 008NF16-KT9R
COMPARE
Red
$134.25USD / Pkg of 10