Diba Dibafit™ Type S Ferrules for Flat-Bottom 1/4"-28 UNF(M) and M6 ISO(M) Fittings
Fits permanently onto the outside of semi-rigid tubing
  • Rated for pressures up to 500 psi (34 bar)
Type S system uses a two-piece ferrule made up of an inner PEEK™ or ETFE sealing surface surrounded by a stainless steel lock ring with only the inner portion in the fluid path.
ITEM(S) ON SALE
$40.80 - $233.50USD / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material
Availability Pricing
Diba Omni-Lok™ Type-S Ferrule, 1/16" OD tubing, PEEK™ / SS, flat bottom; 10/pk EW-21940-49 Mfr # 008FK16
COMPARE
1/16 PEEK ferrule/SS locking ring
$65.40USD / Pkg of 10
Diba Omni-Lok™ Type-S Ferrule, 1/8" OD tubing, PEEK™ / SS, flat bottom; 10/pk EW-21940-55 Mfr # 008FK32
COMPARE
1/8 PEEK ferrule/SS locking ring
$65.40USD / Pkg of 10
Diba Omni-Lok™ Type-S Ferrule, 3/16" OD tubing, PEEK™ / SS, flat bottom; 10/pk EW-21940-56 Mfr # 008FK47
COMPARE
3/16 PEEK ferrule/SS locking ring
$179.75USD / Pkg of 10
Diba Omni-Lok™ Type-S Ferrule, 1/16" OD tubing, ETFE / SS, flat bottom; 10/pk EW-21940-66 Mfr # 008FZ16
COMPARE
1/16 ETFE ferrule/SS locking ring
$58.10USD / Pkg of 10
Diba Omni-Lok™ Type-S Ferrule, 1/8" OD tubing, ETFE / SS, flat bottom; 10/pk EW-21940-67 Mfr # 008FZ32
COMPARE
1/8 ETFE ferrule/SS locking ring
$40.80USD / Pkg of 10
$62.00USD / Pkg of 10
Diba Omni-Lok™ Type-S Ferrule, 3/16" OD tubing, ETFE / SS, flat bottom; 10/pk EW-21940-68 Mfr # 008FZ47
COMPARE
3/16 ETFE ferrule/SS locking ring
$233.50USD / Pkg of 10