Diba Omni-Lok™ 1/4"-28 UNF(M) Fittings for Inverted Cones, 1/8" OD Tubing
Removable and reuable system provides quick and convenient connections
  • ETFE cone is the only material in the fluid path, providing excellent chemical resistance
None of the parts are permanently attached to the tubing, so both fittings and inverted cones can easily be removed and reused. Select polypropylene (PP) compact head fittings from a range of colors for easy identification. PEEK fittings are excellent for higher temperature applications and are available in compact head, standard head, or short thread styles. Compact head fittings are ideal for tight spaces; short thread fittings are great for shallow thread ports.
Select from short or long inverted cones (ferrules). Short inverted cones are designed to give most thread engagement in shallow ports.
$28.20 - $87.60USD / Pkg of 10
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Description
Availability Pricing
Diba Omni-Lok™ Tubing End Fitting, inverted cone, 1/8"OD, 1/4"-28 UNFM; 10/pk EW-21939-94 Mfr # 002244
COMPARE
Natural PEEK Omni-Lok™ Tubing End Fitting, inverted cone, 1/8"OD, 1/4"-28 UNFM; 10/pk
$87.60USD / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, inverted cone, 1/8"OD, 1/4"-28 UNFM; 10/pk EW-21939-95 Mfr # 002245
COMPARE
Natural PEEK Omni-Lok™ Compact Tubing End Fitting, inverted cone, 1/8"OD, 1/4"-28 UNFM; 10/pk
$84.20USD / Pkg of 10
Diba Omni-Lok™ Tubing End Fitting, inverted cone, 1/8"OD tubing, 1/4"-28 UNFM, blue PP; 10/pk EW-21941-85 Mfr # 008NC32-YA5U
COMPARE
Blue Polypropylene Omni-Lok™ Tubing End Fitting, inverted cone, 1/8"OD tubing, 1/4"-28 UNFM, blue PP; 10/pk
$28.20USD / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, inverted cone, 1/8"OD, 1/4"-28 UNFM, green PP; 10/pk EW-21941-86 Mfr # 008NC32-YC5G
COMPARE
Green Polypropylene Omni-Lok™ Compact Tubing End Fitting, inverted cone, 1/8"OD, 1/4"-28 UNFM, green PP; 10/pk
$32.80USD / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, inverted cone, 1/8"OD, 1/4"-28 UNFM, orng PP; 10/pk EW-21941-87 Mfr # 008NC32-YC5N
COMPARE
Orange Polypropylene Omni-Lok™ Compact Tubing End Fitting, inverted cone, 1/8"OD, 1/4"-28 UNFM, orng PP; 10/pk
$32.80USD / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, inverted cone, 1/8"OD, 1/4"-28 UNFM, red PP; 10/pk EW-21941-88 Mfr # 008NC32-YC5R
COMPARE
Red Polypropylene Omni-Lok™ Compact Tubing End Fitting, inverted cone, 1/8"OD, 1/4"-28 UNFM, red PP; 10/pk
$32.80USD / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, inverted cone, 1/8"OD, 1/4"-28 UNFM, blue PP; 10/pk
Always in Stock
EW-21941-89 Mfr # 008NC32-YC5U
COMPARE
Blue Polypropylene Omni-Lok™ Compact Tubing End Fitting, inverted cone, 1/8"OD, 1/4"-28 UNFM, blue PP; 10/pk
$32.80USD / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, inverted cone, 1/8"OD, 1/4"-28 UNFM, yellw PP; 10/pk EW-21941-90 Mfr # 008NC32-YC5Y
COMPARE
Yellow Polypropylene Omni-Lok™ Compact Tubing End Fitting, inverted cone, 1/8"OD, 1/4"-28 UNFM, yellw PP; 10/pk
$32.80USD / Pkg of 10
Showing 1- 8 of 8