Diba Omni-Lok™ 1/4"-28 UNF(M) Fittings for Type P/S Ferrules, 1/16" OD Tubing
Permanently attached ferrules are ideal for repeated connect/disconnect
  • Free spinning fitting eliminates tubing from twisting
  • A recess in the fitting houses the ferrule allowing maximum thread engagement with the port
Type P ferrule has inner PTFE sealing face, the only material in the fluid path. Two-piece type S ferrule consists of an inner PEEK or ETFE sealing face surrounded by a stainless steel lock ring, with only the inner portion in the fluid path. Choose type P for higher pressures, type S for higher temperatures. Select glass-filled polypropylene compact head fittings available in a range of colors for easy identification. Long head fitting is ideal where accessibility is an issue. Use short thread option for most thread engagement in shallow ports.
$28.20 - $87.60USD / Pkg of 10
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Description
Availability Pricing
Diba Omni-Lok™ Short Thread Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), PEEK; 10/pk EW-21940-27 Mfr # 002122S
COMPARE
Natural PEEK Omni-Lok™ Short Thread Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), PEEK; 10/pk
$87.60USD / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), gray PP; 10/pk EW-21942-07 Mfr # 008NF16-YC5A
COMPARE
Gray Polypropylene Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), gray PP; 10/pk
$28.20USD / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), black PP; 10/pk EW-21942-08 Mfr # 008NF16-YC5B
COMPARE
Black Polypropylene Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), black PP; 10/pk
$32.80USD / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), green PP; 10/pk EW-21942-09 Mfr # 008NF16-YC5G
COMPARE
Green Polypropylene Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), green PP; 10/pk
$32.80USD / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), white PP; 10/pk EW-21942-10 Mfr # 008NF16-YC5H
COMPARE
White Polypropylene Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), white PP; 10/pk
$32.80USD / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), mixed PP; 8/pk EW-21942-11 Mfr # 008NF16-YC5M
COMPARE
Mix Polypropylene Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), mixed PP; 8/pk
$32.80USD / Pkg of 8
Diba Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), orange PP; 10/pk EW-21942-12 Mfr # 008NF16-YC5N
COMPARE
Orange Polypropylene Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), orange PP; 10/pk
$32.80USD / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), red PP; 10/pk EW-21942-13 Mfr # 008NF16-YC5R
COMPARE
Red Polypropylene Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), red PP; 10/pk
$32.80USD / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), blue PP; 10/pk EW-21942-14 Mfr # 008NF16-YC5U
COMPARE
Blue Polypropylene Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), blue PP; 10/pk
$32.80USD / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), yellow PP; 10/pk EW-21942-15 Mfr # 008NF16-YC5Y
COMPARE
Yellow Polypropylene Omni-Lok™ Compact Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), yellow PP; 10/pk
$28.20USD / Pkg of 10
Diba Omni-Lok™ Long Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), black PP; 10/pk EW-21942-17 Mfr # 008NF16-YL5B
COMPARE
Black Polypropylene Omni-Lok™ Long Tubing End Fitting, Type S/P, 1/16"OD, 1/4"-28 UNF(M), black PP; 10/pk
$44.40USD / Pkg of 6
Showing 1- 11 of 11