Diba Omni-Lok™ M6 ISO(M) and 5/16"-24 UNF(M) Compact Head Fittings for Inverted Cones, PP
Removable and reuable system provides quick and convenient connections
  • ETFE cone is the only material in the fluid path, providing excellent chemical resistance
  • Compact head fittings are great for tight spaces
None of the parts are permanently attached to the tubing, so both fittings and inverted cones can easily be removed and reused. Select from short or long inverted cones (ferrules). Short inverted cones are designed to give most thread engagement in shallow ports.
$28.20 - $46.60USD / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Operating Pressure (PSI) Connection Type Description
Availability Pricing
Diba Omni-Lok™ Compact Tubing End Fitting, inverted cone, 1/16"OD, M6 ISO(M), blue PP; 10/pk EW-21941-74 Mfr # 008NC16-YC6U
COMPARE
250 Compression x ISO(M) Omni-Lok™ Compact Tubing End Fitting, inverted cone, 1/16"OD, M6 ISO(M), blue PP; 10/pk
$28.20USD / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, inverted cone, 1/8"OD, M6 ISO(M), blue PP; 10/pk EW-21941-91 Mfr # 008NC32-YC6U
COMPARE
250 Compression x ISO(M) Omni-Lok™ Compact Tubing End Fitting, inverted cone, 1/8"OD, M6 ISO(M), blue PP; 10/pk
$28.20USD / Pkg of 10
Diba Omni-Lok™ Compact Tubing End Fitting, inverted cone, 3/16"OD, 5/16"-24 UNFM, blu PP; 10/pk EW-21941-92 Mfr # 008NC47-YC7U
COMPARE
200 Compression x UNF(M) Omni-Lok™ Compact Tubing End Fitting, inverted cone, 3/16"OD, 5/16"-24 UNFM, blu PP; 10/pk
$46.60USD / Pkg of 10