Diba Omnifit® PTFE Cap System Fittings and Valves with 1/4"-28 UNF(M) Connection
Fits into a 1/4"-28 UNF(F) port and accepts 1 to 4 mm OD semi-rigid and rigid tubing
  • Select fittings with on/off valve to control the flow
Quickly and easily connect semi-rigid tubing such as PTFE, ETFE, and FEP as well as rigid materials such as glass, PEEK, and stainless steel. Each fitting includes polypropylene (PP) caps, O-rings/seal, and PTFE cones (1.5 mm OD). Use the interchangeable components to create chemically inert fluid pathway connections and flat-bottom ports (using Gripper fittings). Fittings with integral on/off valve helps control the flow of your fluid line. Fittings are autoclavable (remove caps first). Order colored caps quickly and easily color code multiple fluid lines.
$63.30 - $182.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Number Of Ports Description
Availability Pricing
Diba Omnifit® Cap System Adapter, Tee, 1/4"-28 UNF(M) branch port; 1/ea EW-21939-10 Mfr # 01012
COMPARE
3 Omnifit® Cap System Adapter, Tee, 1/4"-28 UNF(M) branch port; 1/ea
$97.00USD / Each
Diba Omnifit® Cap System Adapter with Column Bleed Valve, Tee, 1/4"-28 UNF(M); 1/ea EW-21939-28 Mfr # 001131
COMPARE
3 Omnifit® Cap System Adapter with Column Bleed Valve, Tee, 1/4"-28 UNF(M); 1/ea
$182.00USD / Each
Diba Omnifit® Cap System Adapter, 1/4"-28 UNF(M); 1/ea EW-21940-15 Mfr # 002506
COMPARE
2 Omnifit® Cap System Adapter, 1/4"-28 UNF(M); 1/ea
$63.30USD / Each
Diba Omnifit® Cap System Adapter with Shut-Off Valve, 1/4"-28 UNF(M); 1/ea EW-21940-16 Mfr # 002507
COMPARE
2 Omnifit® Cap System Adapter with Shut-Off Valve, 1/4"-28 UNF(M); 1/ea
$101.25USD / Each