Dibafit™ Gripper Ferrules
Chemically inert flangeless fitting with permanently attached PTFE ferrule ideal for repeat connect/disconnect
  • Available in PTFE/SS or PTFE/PEEK material
The Dibafit™ Gripper fitting system uses a one-piece ferrule which fits permanently onto the outside of semi-rigid tubing.
$119.00USD / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Material Compatible With
Availability Pricing
Dibafit™ Gripper Ferrule, PTFE/Stainless Steel, 1/4"-28 UNF x 1/16" OD; 10/Pk
Always in Stock
EW-21940-00 Mfr # 2310
COMPARE
$119.00USD / Pkg of 10
Dibafit™ Gripper Ferrule, PTFE/Stainless Steel, 1/4"-28 UNF x 1/8" OD; 10/Pk EW-21940-01 Mfr # 002312
COMPARE
$119.00USD / Pkg of 10
Dibafit™ Gripper Ferrule, PTFE/Stainless Steel, M6 ISO Thread x 1/16" OD; 10/Pk EW-21940-02 Mfr # 002313
COMPARE
$119.00USD / Pkg of 10