Digi-Sense RTD High-Temperature Probes
Ideal for critical process control and heat treating applications
  • Time Constant: 3 seconds or greater
  • Custom designs or changes to this product are available upon request
Alpha coefficient is 0.003850 omega/omega/°C (DIN IEC 751). Resistance at ice point (0°C) is 100 omega.
Note: Time constant is the time required to reach 63.2% of the new reading. To determine how long it will take to reach 99% of the new reading multiply the time constant by five.
.
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
$352.50 - $366.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F)
Availability Pricing
COMPARE
Digi-Sense 3-pin 1450
$366.50USD / Each
COMPARE
5-Pin DIN
$352.50USD / Each