Dwyer Type 475 Manometer

Measures positive, negative and differential pressures
  • ±0.5% full scale accuracy
  • Rugged, extruded aluminum case
  • Selectable english/metric units
The Dwyer® Series 475 Mark III handheld digital manometer is ideal for field calibration, monitoring or trouble shooting HVAC systems, clean rooms or a wide range of other low pressure pneumatic systems. This handy instrument measures positive, negative or differential pressures of air and natural gases in ranges from 1" WC to 150 psi. The manometer is approved and is intrinsically safe for hazardous locations, Class 1, Div.1, Group A, B,C, D, T4. Its simple operation and easy to read digital display make it an indispensable test instrument for the plant engineer, industrial hygienist and HVAC technician. The manometer is housed in a durable extruded aluminum case with its solid state circuitry mounted on a tough fiberglass epoxy circuit board. To meet the most demanding applicat
$333.50 - $660.00USD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Dwyer 475-000-FM Digital Manometer, 1" WC EW-68062-66 Mfr # 475-000-FM
COMPARE
±0.5% full-scale (60 to 78°F); ±1.5% full-scale below 60°F and above 78°F 0.001" WC
$660.00USD / Each
Dwyer 475-0-FM Digital Manometer, 10" WC EW-68062-70 Mfr # 475-0-FM
COMPARE
±0.5% full-scale (60 to 78°F); ±1.5% full-scale below 60°F and above 78°F 0.01" WC
$420.75USD / Each
Dwyer 475-6-FM Digital Manometer, 30 psi EW-68062-76 Mfr # 475-6-FM
COMPARE
±0.5% full-scale (60 to 78°F); ±1.5% full-scale below 60°F and above 78°F 30 0.01 psi
$333.50USD / Each
Dwyer 475-8-FM Digital Manometer, 150 psi EW-68062-80 Mfr # 475-8-FM
COMPARE
±0.5% full-scale (60 to 78°F); ±1.5% full-scale below 60°F and above 78°F 150 0.1 psi
$377.00USD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat