Dynalon Disposable Dropper Pipettes, LDPE
Ergonomic bellows-shaped leakproof bulb reduces finger strain
  • Nominal graduations to indicate volume
One-piece droppers are constructed of low-density polyethylene (LDPE) and are particularly useful for all applications with infectious or toxic substances. Graduated to give an indication of the amount of liquid being drawn. Extremely easy-to use bellows-shaped leakproof bulb provides excellent drop formation.
$35.80 - $60.90USD / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Drops Per mL Graduations (mL) Max Volume (mL)
Availability Pricing
Dynalon Disposable Dropper Pipette, LDPE, 1.5 mL; 100/Pk
Buy More Save More
EW-06223-15 Mfr # 262175
COMPARE
18 0.5 1.5
$60.90USD / Pkg of 100
Dynalon Disposable Dropper Pipette, LDPE, 5.0 mL; 100/Pk
Buy More Save More
EW-06223-25 Mfr # 262185
COMPARE
15 1 5
$35.80USD / Pkg of 100