Dynalon Wide-Mouth, Polypropylene, Graduated Bottles with Write-On Label
Complete with printed white label for clear, easy, and safe labeling!
Manufactured in translucent polypropylene, these bottles are chemical resistant, leakproof, and allow safe storage for a wide variety of liquids and solids. Bottles are wide-mouth for easy filling and have printed graduations in both milliliters and fluid ounces. Screw caps included. Autoclavable.
$62.40 - $160.25USD / Pkg of 12
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Dynalon Graduated Wide-Mouth Bottle, PP, write-on label, 60 mL, 12/pk
Buy More Save More
EW-06041-10 Mfr # 501505-0060
COMPARE
60 10 Polypropylene(PP)
$62.40USD / Pkg of 12
Dynalon Graduated Wide-Mouth Bottle, PP, write-on label, 125 mL, 12/pk
Buy More Save More
EW-06041-12 Mfr # 501505-0125
COMPARE
125 20 Polypropylene(PP)
$65.00USD / Pkg of 12
Dynalon Graduated Wide-Mouth Bottle, PP, write-on label, 250 mL, 12/pk
Buy More Save More
EW-06041-14 Mfr # 501505-0250
COMPARE
250 50 Polypropylene(PP)
$81.50USD / Pkg of 12
Dynalon Graduated Wide-Mouth Bottle, PP, write-on label, 500 mL, 12/pk
Buy More Save More
EW-06041-16 Mfr # 501505-0500
COMPARE
500 100 Polypropylene(PP)
$108.75USD / Pkg of 12
Dynalon Graduated Wide-Mouth Bottle, PP, write-on label, 1 L, 6/pk
Buy More Save More
EW-06041-18 Mfr # 501505-1000
COMPARE
1000 100 Polypropylene(PP)
$160.25USD / Pkg of 6