Edwards Inlet/Outlet Filter for XDS and nXDS Dry Scroll Vacuum Pumps
Place on inlet or outlet to protect your vacuum pump
  • Supplied ready to use with a 5 µm filtering element
Dual-purpose filter can be placed at the inlet or the outlet of your pump. Place between the pump inlet and your chamber outlet connection to prevent dust from entering into your system. Place it at the outlet between the vacuum pump and the exhaust line to trap out tip-seal dust.
$1,175.00 - $1,475.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Connections Compatible With
Availability Pricing
Edwards Inlet/Exhaust Filter NW25 for XDS and nXDS Dry Scroll Vacuum Pumps, 5 µm EW-79301-41 Mfr # A50597805
COMPARE
NW25 XDS and nXDS dry scroll vacuum pumps with NW25 port
$1,175.00USD / Each
Edwards Inlet/Exhaust Filter NW40 for XDS Dry Scroll Vacuum Pumps, 5 µm EW-79301-68 Mfr # A50597806
COMPARE
NW40 XDS dry scroll vacuum pumps with NW40 port
$1,475.00USD / Each