Edwards nXDS Dry Scroll Pumps
Exceptional pumping capability with oil-free operation
  • Prevent contamination of media and extend bearing life with bearing shields
  • Advanced controller allows for remote control
  • Wisper quiet operation for minimal interruption
These vacuum scroll pumps are an ideal alternative where oil-free pumping is required. Oilless pump operation eliminates the cost and downtime associated with oil changes and oil disposal. The nXDS series pumps feature a bearing shield to isolate the vacuum environment from all forms of lubricant not only making them totally dry, but also protecting the bearing from any process gases. nXDS improves on legacy XDS pumps by offering increased pumping speeds, combined with lower ultimate pressures, lower power consumption, and lower noise.
Operate the pump remotely with the advanced interface providing a variety of traditional and analogue and digital control methods enabling remote control/monitoring via RS-232 or RS-485 communication. The USB port is included for service use only. These vacuum pumps are designed to be completely field serviceable, and the unique tip seal design significantly reduces wear to minimize maintenance and extend service life.
Select pump models are available without gas ballast for applications such as gas recovery. The chemical versions feature perfluoroelastomer internal valves and stainless steel fittings for enhanced protection from corrosive media.
$9,920.00 - $17,700.00USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free Air Capacity (CFM) Max Vacuum (torr) Description
Availability Pricing
Edwards nXDS6i Dry Scroll Vacuum Pump; 4 CFM EW-79301-19 Mfr # A73501983
COMPARE
4 0.015 nXDS6i Dry Scroll Vacuum Pump; 4 CFM
$9,920.00USD / Each
Edwards nXDS6iC Dry Scroll Vacuum Pump, Chemical Resistant; 4 CFM EW-79301-22 Mfr # A73502983
COMPARE
4 0.015 nXDS6iC Dry Scroll Vacuum Pump, Chemical Resistant; 4 CFM
$10,865.00USD / Each
Edwards nXDS10i Dry Scroll Vacuum Pump; 7.5 CFM EW-79301-24 Mfr # A73601983
COMPARE
7.5 0.007 nXDS10i Dry Scroll Vacuum Pump; 7.5 CFM
$11,255.00USD / Each
Edwards nXDS10iC Dry Scroll Vacuum Pump, Chemical Resistant; 7.5 CFM EW-79301-25 Mfr # A73602983
COMPARE
7.5 0.007 nXDS10iC Dry Scroll Vacuum Pump, Chemical Resistant; 7.5 CFM
$13,750.00USD / Each
Edwards nXDS10iR Dry Scroll Vacuum Pump, No Gas Ballast; 7.5 CFM EW-79301-26 Mfr # A73603983
COMPARE
7.5 0.007 nXDS10iR Dry Scroll Vacuum Pump, No Gas Ballast; 7.5 CFM
$11,635.00USD / Each
Edwards nXDS15i Dry Scroll Vacuum Pump; 10.1 CFM EW-79301-27 Mfr # A73701983
COMPARE
10.1 0.007 nXDS15i Dry Scroll Vacuum Pump; 10.1 CFM
$14,020.00USD / Each
Edwards nXDS15iC Dry Scroll Vacuum Pump, Chemical Resistant; 10.1 CFM EW-79301-28 Mfr # A73702983
COMPARE
10.1 0.007 nXDS15iC Dry Scroll Vacuum Pump, Chemical Resistant; 10.1 CFM
$15,025.00USD / Each
Edwards nXDS20i Dry Scroll Vacuum Pump; 16.5 CFM EW-79301-30 Mfr # A73801983
COMPARE
16.5 0.022 nXDS20i Dry Scroll Vacuum Pump; 16.5 CFM
$16,040.00USD / Each
Edwards nXDS20iC Dry Scroll Vacuum Pump, Chemical Resistant; 16.5 CFM EW-79301-31 Mfr # A73802983
COMPARE
16.5 0.022 nXDS20iC Dry Scroll Vacuum Pump, Chemical Resistant; 16.5 CFM
$17,700.00USD / Each
Showing 1- 9 of 9