Edwards XDS Dry Scroll Pumps
Compact pump size with robust pumping speed
  • Optimize performance with safe motor drive overload protection
  • Prevent contamination of media and extend bearing life with bearing shields
  • No more oil—runs dry!
These vacuum scroll pumps are an ideal alternative where oil-free pumping is required. Oilless pump operation eliminates the cost and downtime associated with oil changes and oil disposal. The XDS series pumps feature a bearing shield to isolate the vacuum environment from all forms of lubricant not only making them totally dry, but also protecting the bearing from any process gases.
The single-sided scroll pump design allows maintenance to be done in minutes. The smart motor drive technology provides pump overload protection and remote start/stop capabilities, allowing for reliable performance. Operate the pump remotely using the 15-way D type connector, located on the side panel of the pump for easy access.
Enhanced models reduced peak power requirements, meaning they can pump down large volume changers with no loss of performance.
Choose from two series—the XDS35i and the XDS46i. Both share many of the same features, however the XDS46i series has a higher free-air capacity and max vacuum.
Select pump models are available without gas ballast for applications such as gas recovery. The chemical versions feature Chemraz® internal valves and stainless steel fittings for enhanced protection from corrosive media.
$20,685.00 - $23,213.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free Air Capacity (CFM) Max Vacuum (torr) Description
Availability Pricing
Edwards XDS35i Dry Scroll Vacuum Pump EW-79301-33 Mfr # A73001983
COMPARE
21 0.008 XDS35i Dry Scroll Vacuum Pump
$20,685.00USD / Each
Edwards XDS35iE Dry Scroll Vacuum Pump, Enhanced EW-79301-36 Mfr # A73003983
COMPARE
21 0.02 XDS35iE Dry Scroll Vacuum Pump, Enhanced
$22,052.00USD / Each
Edwards XDS46i Dry Scroll Vacuum Pump EW-79301-39 Mfr # A73101983
COMPARE
23.5 0.04 XDS46i Dry Scroll Vacuum Pump
$23,213.00USD / Each