Eldon James Hose Barb Fittings, 90° Elbow, Animal-Derivative-Free Polypropylene
Easily change direction of fluid flow with 90° elbows
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. The elbows feature a 90° bend and is designed to fit in a tight space. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
These animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. Not recommended for use with oxidizing agents.
$12.10 - $20.10USD / Pkg of 10
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Connection Type
Availability Pricing
Eldon James Hose Barb Fitting, 90° Elbow, Animal-Derivative-Free Polypropylene, 3/32" ID; 10/Pk EW-50126-69 Mfr # L0-1.5-200PP
COMPARE
3/32 3/32 Hose Barb Elbow
$12.10USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Animal-Derivative-Free Polypropylene, 5/8" ID; 10/Pk EW-50126-70 Mfr # L0-10PP
COMPARE
5/8 5/8 Hose Barb Elbow
$14.90USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Animal-Derivative-Free Polypropylene, 1/16" ID; 10/Pk EW-50126-71 Mfr # L0-1-200PP
COMPARE
1/16 1/16 Hose Barb Elbow
$12.10USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Animal-Derivative-Free Polypropylene, 3/4" ID; 10/Pk EW-50126-72 Mfr # L0-12PP
COMPARE
3/4 3/4 Hose Barb Elbow
$17.20USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Animal-Derivative-Free Polypropylene, 1" ID; 10/Pk EW-50126-73 Mfr # L0-16PP
COMPARE
1 1 Hose Barb Elbow
$20.10USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Animal-Derivative-Free Polypropylene, 5/32" ID; 10/Pk EW-50126-74 Mfr # L0-2.5PP
COMPARE
5/32 5/32 Hose Barb Elbow
$19.60USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Animal-Derivative-Free Polypropylene, 1/8" ID; 10/Pk EW-50126-75 Mfr # L0-2-200PP
COMPARE
1/8 1/8 Hose Barb Elbow
$13.20USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Animal-Derivative-Free Polypropylene, 3/16" ID; 10/Pk EW-50126-76 Mfr # L0-3PP
COMPARE
3/16 3/16 Hose Barb Elbow
$19.60USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Animal-Derivative-Free Polypropylene, 1/4" ID; 10/Pk EW-50126-77 Mfr # L0-4PP
COMPARE
1/4 1/4 Hose Barb Elbow
$15.50USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Animal-Derivative-Free Polypropylene, 5/16" ID; 10/Pk EW-50126-78 Mfr # L0-5PP
COMPARE
5/16 5/16 Hose Barb Elbow
$13.20USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Animal-Derivative-Free Polypropylene, 3/8" ID; 10/Pk EW-50126-79 Mfr # L0-6PP
COMPARE
3/8 3/8 Hose Barb Elbow
$13.20USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Animal-Derivative-Free Polypropylene, 1/2" ID; 10/Pk EW-50126-80 Mfr # L0-8PP
COMPARE
1/2 1/2 Hose Barb Elbow
$13.20USD / Pkg of 10
Showing 1- 12 of 12