Eldon James Hose Barb Fittings, 90° Elbow, Natural Kynar®
Easily change the direction of the fluid flow with 90° elbows
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • Biopharma compatible and USP 661 and BPOG compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. The elbows feature a 90° bend and is designed to fit in a tight space. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability.
$17.20 - $73.20USD / Pkg of 10
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Connection Type
Availability Pricing
Eldon James Hose Barb Fitting, 90° Elbow, Natural Kynar®, 3/32" ID; 10/Pk EW-50128-02 Mfr # L0-1.5-200NK7
COMPARE
3/32 3/32 Hose Barb Elbow
$18.30USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Natural Kynar®, 5/8" ID; 10/Pk EW-50128-03 Mfr # L0-10NK7
COMPARE
5/8 5/8 Hose Barb Elbow
$48.90USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Natural Kynar®, 1/16" ID; 10/Pk EW-50128-04 Mfr # L0-1-200NK7
COMPARE
1/16 1/16 Hose Barb Elbow
$17.20USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Natural Kynar®, 3/4" ID; 10/Pk EW-50128-05 Mfr # L0-12NK7
COMPARE
3/4 3/4 Hose Barb Elbow
$59.70USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Natural Kynar®, 1" ID; 10/Pk EW-50128-06 Mfr # L0-16NK7
COMPARE
1 1 Hose Barb Elbow
$73.20USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Natural Kynar®, 5/32" ID; 10/Pk EW-50128-07 Mfr # L0-2.5NK7
COMPARE
5/32 5/32 Hose Barb Elbow
$32.00USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Natural Kynar®, 1/8" ID; 10/Pk EW-50128-08 Mfr # L0-2-200NK7
COMPARE
1/8 1/8 Hose Barb Elbow
$19.60USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Natural Kynar®, 3/16" ID; 10/Pk EW-50128-09 Mfr # L0-3NK7
COMPARE
3/16 3/16 Hose Barb Elbow
$34.20USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Natural Kynar®, 1/4" ID; 10/Pk EW-50128-10 Mfr # L0-4NK7
COMPARE
1/4 1/4 Hose Barb Elbow
$34.20USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Natural Kynar®, 5/16" ID; 10/Pk EW-50128-11 Mfr # L0-5NK7
COMPARE
5/16 5/16 Hose Barb Elbow
$36.50USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Natural Kynar®, 3/8" ID; 10/Pk EW-50128-12 Mfr # L0-6NK7
COMPARE
3/8 3/8 Hose Barb Elbow
$35.30USD / Pkg of 10
Eldon James Hose Barb Fitting, 90° Elbow, Natural Kynar®, 1/2" ID; 10/PK EW-50128-13 Mfr # L0-8NK7
COMPARE
1/2 1/2 Hose Barb Elbow
$37.60USD / Pkg of 10
Showing 1- 12 of 12