Eldon James Hose Barb Fittings, Straight Reducer, Animal-Derivative-Free Polypropylene
Single barb design tightly “grips” tubing
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a
  • RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Easily connect two different size tubing with straight reducers. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
These animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. However, it’s important to avoid using them with oxidizing agents.
$9.30 - $18.30USD / Pkg of 10
35 variations of this product are available.
See All Product Options Below
35 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 3/32" ID x 1/16" ID; 10/PK EW-50126-34 Mfr # C1.5-1-200PP
COMPARE
3/32 1/16 Polypropylene
$9.30USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 5/8" ID x 1/4" ID; 10/PK EW-50126-35 Mfr # C10-4PP
COMPARE
5/8 1/4 Polypropylene
$13.70USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 5/8" ID x 5/16" ID; 10/PK EW-50126-36 Mfr # C10-5PP
COMPARE
5/8 5/16 Polypropylene
$13.70USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 5/8" ID x 3/8" ID; 10/PK EW-50126-37 Mfr # C10-6PP
COMPARE
5/8 3/8 Polypropylene
$13.70USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 5/8" ID x 1/2" ID; 10/PK EW-50126-38 Mfr # C10-8PP
COMPARE
5/8 1/2 Polypropylene
$13.70USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 3/4" ID x 5/8" ID; 10/PK EW-50126-39 Mfr # C12-10PP
COMPARE
3/4 5/8 Polypropylene
$16.10USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 3/4" ID x 3/8" ID; 10/PK EW-50126-40 Mfr # C12-6PP
COMPARE
3/4 3/8 Polypropylene
$15.50USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 3/4" ID x 1/2" ID; 10/PK EW-50126-41 Mfr # C12-8PP
COMPARE
3/4 1/2 Polypropylene
$13.70USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 1" ID x 5/8" ID; 10/PK EW-50126-42 Mfr # C16-10PP
COMPARE
1 5/8 Polypropylene
$17.80USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 1" ID x 3/4" ID; 10/PK EW-50126-43 Mfr # C16-12PP
COMPARE
1 3/4 Polypropylene
$17.80USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 1" ID x 1/2" ID; 10/PK EW-50126-44 Mfr # C16-8PP
COMPARE
1 1/2 Polypropylene
$17.80USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 5/32" ID x 3/32" ID; 10/PK EW-50126-45 Mfr # C2.5-1.5-200PP
COMPARE
5/32 3/32 Polypropylene
$12.10USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 5/32" ID x 1/8" ID; 10/PK EW-50126-46 Mfr # C2.5-2PP
COMPARE
5/32 1/8 Polypropylene
$13.70USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 1/8" ID x 3/32" ID; 10/PK EW-50126-47 Mfr # C2-1.5-200PP
COMPARE
1/8 3/32 Polypropylene
$9.30USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 1/8" ID x 1/16" ID; 10/PK EW-50126-48 Mfr # C2-1-200PP
COMPARE
1/8 1/16 Polypropylene
$9.30USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 3/16" ID x 3/32" ID; 10/PK EW-50126-49 Mfr # C3-1.5-200PP
COMPARE
3/16 3/32 Polypropylene
$12.10USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 3/16" ID x 1/16" ID; 10/PK EW-50126-50 Mfr # C3-1-200PP
COMPARE
3/16 1/16 Polypropylene
$12.10USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 3/16" ID x 5/32" ID; 10/PK EW-50126-51 Mfr # C3-2.5-200PP
COMPARE
3/16 5/32 Polypropylene
$12.10USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 3/16" ID x 1/8" ID; 10/PK EW-50126-52 Mfr # C3-2PP
COMPARE
3/16 1/8 Polypropylene
$13.70USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Animal-Derivative-Free Polypropylene, 1/4" ID x 3/32" ID; 10/PK EW-50126-53 Mfr # C4-1.5-200PP
COMPARE
1/4 3/32 Polypropylene
$18.30USD / Pkg of 10
Showing 1 - 20 of 35