Eldon James Hose Barb Fittings, Straight Reducer, Stainless Steel
Excellent for high temperature and harsh physical environments
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
These durable stainless steel fittings perform well in high temperatures and challenging environments. Made of 316L molybdenum bearing stainless steel, they are widely used in pharmaceutical, food, and beverage applications. Use the straight reducers to connect two different size tubing. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
$13.20 - $98.20USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
Eldon James Hose Barb Fitting, Straight Reducer, Stainless Steel, 5/32" ID x 3/32" ID; 10/Pk EW-50108-71 Mfr # C2.5-1.5SS/10PK
COMPARE
5/32 3/32 Stainless Steel
$98.20USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Stainless Steel, 1/8" ID x 3/32" ID; 10/Pk EW-50108-78 Mfr # C2-1.5SS/1OPK
COMPARE
1/8 3/32 Stainless Steel
$89.80USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Reducer, Stainless Steel, 5/8" ID x 1/2" ID; 1/Each EW-50127-69 Mfr # C10-8SS
COMPARE
5/8 1/2 Stainless Steel
$41.00USD / Each
Eldon James Hose Barb Fitting, Straight Reducer, Stainless Steel, 3/4" ID x 1/2" ID; 1/Each EW-50127-70 Mfr # C12-8SS
COMPARE
3/4 1/2 Stainless Steel
$27.10USD / Each
Eldon James Hose Barb Fitting, Straight Reducer, Stainless Steel, 5/32" ID x 3/32" ID; 1/Each EW-50127-71 Mfr # C2.5-1.5SS
COMPARE
5/32 3/32 Stainless Steel
$14.10USD / Each
Eldon James Hose Barb Fitting, Straight Reducer, Stainless Steel, 1/8" ID x 3/32" ID; 1/Each EW-50127-72 Mfr # C2-1.5SS
COMPARE
1/8 3/32 Stainless Steel
$13.20USD / Each
Eldon James Hose Barb Fitting, Straight Reducer, Stainless Steel, 1/4" ID x 5/32" ID; 1/Each EW-50127-73 Mfr # C4-2.5SS
COMPARE
1/4 5/32 Stainless Steel
$16.10USD / Each
Eldon James Hose Barb Fitting, Straight Reducer, Stainless Steel, 1/4" ID x 1/8" ID; 1/Each EW-50127-74 Mfr # C4-2SS
COMPARE
1/4 1/8 Stainless Steel
$14.70USD / Each
Eldon James Hose Barb Fitting, Straight Reducer, Stainless Steel, 1/4" ID x 3/16" ID; 1/Each EW-50127-75 Mfr # C4-3SS
COMPARE
1/4 3/16 Stainless Steel
$17.50USD / Each
Eldon James Hose Barb Fitting, Straight Reducer, Stainless Steel, 5/16" ID x 1/4" ID; 1/Each EW-50127-76 Mfr # C5-4SS
COMPARE
5/16 1/4 Stainless Steel
$19.60USD / Each
Eldon James Hose Barb Fitting, Straight Reducer, Stainless Steel, 3/8" ID x 1/4" ID; 1/Each EW-50127-77 Mfr # C6-4SS
COMPARE
3/8 1/4 Stainless Steel
$19.60USD / Each
Eldon James Hose Barb Fitting, Straight Reducer, Stainless Steel, 1/2" ID x 1/4" ID; 1/Each EW-50127-78 Mfr # C8-4SS
COMPARE
1/2 1/4 Stainless Steel
$24.90USD / Each
Showing 1- 12 of 12