Eldon James Hose Barb Fittings, Straight Union, Animal-Derivative-Free Polypropylene
Single barb design tightly “grips” tubing
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose use applications. Easily connect two tubing with straight unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
These animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. Not recommended for use with oxidizing agents.
$9.30 - $18.30USD / Pkg of 10
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
Eldon James Hose Barb Fitting, Straight Union, Animal-Derivative-Free Polypropylene, 3/32" ID; 10/PK EW-50124-76 Mfr # C0-1.5-200PP
COMPARE
3/32 3/32 Polypropylene
$9.30USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Animal-Derivative-Free Polypropylene, 5/8" ID; 10/PK EW-50124-77 Mfr # C0-10PP
COMPARE
5/8 5/8 Polypropylene
$10.90USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Animal-Derivative-Free Polypropylene, 1/16" ID; 10/PK EW-50124-78 Mfr # C0-1-200PP
COMPARE
1/16 1/16 Polypropylene
$9.30USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Animal-Derivative-Free Polypropylene, 3/4" ID; 10/PK EW-50124-79 Mfr # C0-12PP
COMPARE
3/4 3/4 Polypropylene
$16.10USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Animal-Derivative-Free Polypropylene, 1" ID; 10/PK EW-50124-80 Mfr # C0-16PP
COMPARE
1 1 Polypropylene
$18.30USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Animal-Derivative-Free Polypropylene, 5/32" ID; 10/PK EW-50124-81 Mfr # C0-2.5PP
COMPARE
5/32 5/32 Polypropylene
$13.70USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Animal-Derivative-Free Polypropylene, 1/8" ID; 10/PK EW-50124-82 Mfr # C0-2-200PP
COMPARE
1/8 1/8 Polypropylene
$10.90USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Animal-Derivative-Free Polypropylene, 3/16" ID; 10/PK EW-50124-83 Mfr # C0-3PP
COMPARE
3/16 3/16 Polypropylene
$13.70USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Animal-Derivative-Free Polypropylene, 1/4" ID; 10/PK EW-50124-84 Mfr # C0-4PP
COMPARE
1/4 1/4 Polypropylene
$10.30USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Animal-Derivative-Free Polypropylene, 5/16" ID; 10/PK EW-50124-85 Mfr # C0-5PP
COMPARE
5/16 5/16 Polypropylene
$13.70USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Animal-Derivative-Free Polypropylene, 3/8" ID; 10/PK EW-50124-86 Mfr # C0-6PP
COMPARE
3/8 3/8
$10.30USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Animal-Derivative-Free Polypropylene, 1/2" ID; 10/PK EW-50124-87 Mfr # C0-8PP
COMPARE
1/2 1/2 Polypropylene
$15.50USD / Pkg of 10
Showing 1- 12 of 12