Eldon James Hose Barb Fittings, Straight Union, Natural Nylon
Single barb design tightly “grips” tubing
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Easily connect two tubing with straight unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
These natural nylon fittings provide good tensile strength, stiffness, and impact resistance and are resistant to fuels and lubricants. Not recommended for use with phenols, strong acids, and oxidizing agents.
$7.80 - $19.60USD / Pkg of 10
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Fitting Material
Availability Pricing
Eldon James Hose Barb Fitting, Straight Union, Natural Nylon, 3/32" ID; 10/PK EW-50124-64 Mfr # C0-1.5-200NN
COMPARE
3/32 3/32 Nylon
$7.80USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Natural Nylon, 5/8" ID; 10/PK EW-50124-65 Mfr # C0-10NN
COMPARE
5/8 5/8 Nylon
$11.50USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Natural Nylon, 1/16" ID; 10/PK EW-50124-66 Mfr # C0-1-200NN
COMPARE
1/16 1/16 Nylon
$7.80USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Natural Nylon, 3/4" ID; 10/PK EW-50124-67 Mfr # C0-12NN
COMPARE
3/4 3/4 Nylon
$18.30USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Natural Nylon, 1" ID; 10/PK EW-50124-68 Mfr # C0-16NN
COMPARE
1 1 Nylon
$19.60USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Natural Nylon, 5/32" ID; 10/PK EW-50124-69 Mfr # C0-2.5NN
COMPARE
5/32 5/32 Nylon
$13.20USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Natural Nylon, 1/8" ID; 10/PK EW-50124-70 Mfr # C0-2-200NN
COMPARE
1/8 1/8 Nylon
$9.30USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Natural Nylon, 3/16" ID; 10/PK EW-50124-71 Mfr # C0-3NN
COMPARE
3/16 3/16 Nylon
$13.20USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Natural Nylon, 1/4" ID; 10/PK EW-50124-72 Mfr # C0-4NN
COMPARE
1/4 1/4 Nylon
$10.90USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Natural Nylon, 5/16" ID; 10/PK EW-50124-73 Mfr # C0-5NN
COMPARE
5/16 5/16 Nylon
$11.50USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Natural Nylon, 3/8" ID; 10/PK EW-50124-74 Mfr # C0-6NN
COMPARE
3/8 3/8 Nylon
$10.90USD / Pkg of 10
Eldon James Hose Barb Fitting, Straight Union, Natural Nylon, 1/2" ID; 10/PK EW-50124-75 Mfr # C0-8NN
COMPARE
1/2 1/2 Nylon
$13.20USD / Pkg of 10
Showing 1- 12 of 12