Eldon James Hose Barb Fittings, Straight Union, Stainless Steel
Excellent for high temperature and harsh physical environments
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
Easily connect two tubing with straight unions. These durable stainless steel fittings perform well in high temperatures and challenging environments. Made of 316L molybdenum bearing stainless steel, they are widely used in pharmaceutical, food, and beverage applications. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
$10.10 - $58.60USD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Connection Type
Availability Pricing
Eldon James Hose Barb Fitting, Straight Union, Stainless Steel, 5/8" ID; 1/Each EW-50124-88 Mfr # C0-10SS
COMPARE
5/8 5/8 Hose Barb Union
$41.60USD / Each
Eldon James Hose Barb Fitting, Straight Union, Stainless Steel, 3/4" ID; 1/Each EW-50124-89 Mfr # C0-12SS
COMPARE
3/4 3/4 Hose Barb Union
$58.60USD / Each
Eldon James Hose Barb Fitting, Straight Union, Stainless Steel, 5/32" ID; 1/Each EW-50124-90 Mfr # C0-2.5SS
COMPARE
5/32 5/32 Hose Barb Union
$10.90USD / Each
Eldon James Hose Barb Fitting, Straight Union, Stainless Steel, 1/8" ID; 1/Each EW-50124-91 Mfr # C0-2SS
COMPARE
1/8 1/8 Hose Barb Union
$10.10USD / Each
Eldon James Hose Barb Fitting, Straight Union, Stainless Steel, 3/16" ID; 1/Each EW-50124-92 Mfr # C0-3SS
COMPARE
3/16 3/16 Hose Barb Union
$11.80USD / Each
Eldon James Hose Barb Fitting, Straight Union, Stainless Steel, 1/4" ID; 1/Each EW-50124-93 Mfr # C0-4SS
COMPARE
1/4 1/4 Hose Barb Union
$13.50USD / Each
Eldon James Hose Barb Fitting, Straight Union, Stainless Steel, 5/16" ID; 1/Each EW-50124-94 Mfr # C0-5SS
COMPARE
5/16 5/16 Hose Barb Union
$14.90USD / Each
Eldon James Hose Barb Fitting, Straight Union, Stainless Steel, 3/8" ID; 1/Each EW-50124-95 Mfr # C0-6SS
COMPARE
3/8 3/8 Hose Barb Union
$23.20USD / Each
Eldon James Hose Barb Fitting, Straight Union, Stainless Steel, 1/2" ID; 1/Each EW-50124-96 Mfr # C0-8SS
COMPARE
1/2 1/2 Hose Barb Union
$34.20USD / Each
Showing 1- 9 of 9