Eldon James Hose Barb Fittings, Tee Union, Animal-Derivative-Free Polypropylene
Single barb design tightly “grips” tubing
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a
  • RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Easily combine or split fluid flow using tee unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
These animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. However, it’s important to avoid using them with oxidizing agents, as these can compromise their integrity.
$10.90 - $19.60USD / Pkg of 10
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in)
Availability Pricing
Eldon James Hose Barb Fitting, Tee Union, Animal-Derivative-Free Polypropylene, 3/32" ID; 10/PK EW-50127-44 Mfr # T0-1.5-200PP
COMPARE
3/32 3/32 3/32
$13.20USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Animal-Derivative-Free Polypropylene, 5/8" ID; 10/PK EW-50127-45 Mfr # T0-10PP
COMPARE
5/8 5/8 5/8
$14.90USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Animal-Derivative-Free Polypropylene, 1/16" ID; 10/PK EW-50127-46 Mfr # T0-1-200PP
COMPARE
1/16 1/16 1/16
$12.10USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Animal-Derivative-Free Polypropylene, 3/4" ID; 10/PK EW-50127-47 Mfr # T0-12PP
COMPARE
3/4 3/4 3/4
$15.50USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Animal-Derivative-Free Polypropylene, 1" ID; 10/PK EW-50127-48 Mfr # T0-16PP
COMPARE
1 1 1
$18.30USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Animal-Derivative-Free Polypropylene, 5/32" ID; 10/PK EW-50127-49 Mfr # T0-2.5PP
COMPARE
5/32 5/32 5/32
$19.60USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Animal-Derivative-Free Polypropylene, 1/8" ID; 10/PK EW-50127-50 Mfr # T0-2-200PP
COMPARE
1/8 1/8 1/8
$10.90USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Animal-Derivative-Free Polypropylene, 3/16" ID; 10/PK EW-50127-51 Mfr # T0-3PP
COMPARE
3/16 3/16 3/16
$13.20USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Animal-Derivative-Free Polypropylene, 1/4" ID; 10/PK EW-50127-52 Mfr # T0-4PP
COMPARE
1/4 1/4 1/4
$13.20USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Animal-Derivative-Free Polypropylene, 5/16" ID; 10/PK EW-50127-53 Mfr # T0-5PP
COMPARE
5/16 5/16 5/16
$14.90USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Animal-Derivative-Free Polypropylene, 3/8" ID; 10/PK EW-50127-55 Mfr # T0-6PP
COMPARE
3/8 3/8 3/8
$13.20USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Animal-Derivative-Free Polypropylene, 1/2" ID; 10/PK EW-50127-56 Mfr # T0-8PP
COMPARE
1/2 1/2 1/2
$17.20USD / Pkg of 10
Showing 1- 12 of 12