Eldon James Hose Barb Fittings, Tee Union, Natural Kynar®
Single barb design tightly “grips” tubing
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Biopharma compatible and USP 661
  • BPOG compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Easily combine or split fluid flow using tee unions. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability.
$20.10 - $83.40USD / Pkg of 10
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in)
Availability Pricing
Eldon James Hose Barb Fitting, Tee Union, Natural Kynar®, 3/32" ID; 10/PK EW-50128-80 Mfr # T0-1.5-200NK7
COMPARE
3/32 3/32 3/32
$21.10USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Natural Kynar®, 5/8" ID; 10/PK EW-50128-81 Mfr # T0-10NK7
COMPARE
5/8 5/8 5/8
$54.10USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Natural Kynar®, 1/16" ID; 10/PK EW-50128-82 Mfr # T0-1-200NK7
COMPARE
1/16 1/16 1/16
$20.10USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Natural Kynar®, 3/4" ID; 10/PK EW-50128-83 Mfr # T0-12NK7
COMPARE
3/4 3/4 3/4
$66.50USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Natural Kynar®, 1" ID; 10/PK EW-50128-84 Mfr # T0-16NK7
COMPARE
1 1 1
$83.40USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Natural Kynar®, 5/32" ID; 10/PK EW-50128-85 Mfr # T0-2.5NK7
COMPARE
5/32 5/32 5/32
$32.00USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Natural Kynar®, 1/8" ID; 10/PK EW-50128-86 Mfr # T0-2-200NK7
COMPARE
1/8 1/8 1/8
$24.00USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Natural Kynar®, 3/16" ID; 10/PK EW-50128-87 Mfr # T0-3NK7
COMPARE
3/16 3/16 3/16
$37.60USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Natural Kynar®, 1/4" ID; 10/PK EW-50128-88 Mfr # T0-4NK7
COMPARE
1/4 1/4 1/4
$34.20USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Natural Kynar®, 5/16" ID; 10/PK EW-50128-89 Mfr # T0-5NK7
COMPARE
5/16 5/16 5/16
$37.60USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Natural Kynar®, 3/8" ID; 10/PK EW-50128-90 Mfr # T0-6NK7
COMPARE
3/8 3/8 3/8
$37.60USD / Pkg of 10
Eldon James Hose Barb Fitting, Tee Union, Natural Kynar®, 1/2" ID; 10/PK EW-50128-91 Mfr # T0-8NK7
COMPARE
1/2 1/2 1/2
$42.20USD / Pkg of 10
Showing 1- 12 of 12