Eldon James Hose Barb Fittings, Y Reducer, Natural Kynar®
Single barb design tightly “grips” tubing
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Biopharma compatible and USP 661
  • BPOG compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. They allow for easy fluid flow combination or splitting with reduction Y fittings, which have one large hose barb and two smaller ones. Each fitting features a single barb on each end, ensuring a tight seal to keep the tubing securely in place.
An anti-rotation device on each port prevents tubing from rotating, minimizing kinks. These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability.
$44.50 - $209.50USD / Pkg of 10
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in)
Availability Pricing
Eldon James Hose Barb Fitting, Y Reducer, Natural Kynar®, 5/8" ID x 3/4" ID; 10/PK EW-50128-73 Mfr # RY10-12NK7
COMPARE
3/4 5/8 5/8
$191.50USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Reducer, Natural Kynar®, 3/4" ID x 1" ID; 10/PK EW-50128-74 Mfr # RY12-16NK7
COMPARE
1 3/4 3/4
$209.50USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Reducer, Natural Kynar®, 1/8" ID x 1/4" ID; 10/PK EW-50128-75 Mfr # RY2-4NK7
COMPARE
1/40 1/8 1/8
$44.50USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Reducer, Natural Kynar®, 3/16" ID x 5/16" ID; 10/PK EW-50128-76 Mfr # RY3-5NK7
COMPARE
5/16 3/16 3/16
$44.50USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Reducer, Natural Kynar®, 1/4" ID x 3/8" ID; 10/PK EW-50128-77 Mfr # RY4-6NK7
COMPARE
3/8 1/4 1/4
$103.75USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Reducer, Natural Kynar®, 3/8" ID x 1/2" ID; 10/PK EW-50128-78 Mfr # RY6-8NK7
COMPARE
1/2 3/8 3/8
$117.25USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Reducer, Natural Kynar®, 1/2" ID x 5/8" ID; 10/PK EW-50128-79 Mfr # RY8-10NK7
COMPARE
5/8 1/2 1/2
$154.50USD / Pkg of 10
Showing 1- 7 of 7