Eldon James Hose Barb Fittings, Y Union, Natural Nylon
Single barb design tightly “grips” tubing
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose use applications. Easily combine or split fluid flow using Y unions. Each fitting features a single barb on each end, ensuring a tight seal to keep the tubing securely in place.
An anti-rotation device on each port prevents tubing from rotating, minimizing kinks. These natural nylon fittings offer excellent tensile strength, stiffness, and impact resistance, making them suitable for various applications. Their resistance to fuels and lubricants is a definite advantage in many industrial settings. However, it’s important to avoid using them with phenols, strong acids, and oxidizing agents, as these can compromise their integrity.
$12.70 - $47.90USD / Pkg of 10
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Barb Size / Tubing Id (Port 3) (in)
Availability Pricing
Eldon James Hose Barb Fitting, Y Union, Natural Nylon, 3/32" ID; 10/PK EW-50126-22 Mfr # Y0-1.5NN
COMPARE
3/32 3/32 3/32
$12.70USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Nylon, 5/8" ID; 10/PK EW-50126-23 Mfr # Y0-10NN
COMPARE
5/8 5/8 5/8
$37.60USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Nylon, 3/4" ID; 10/PK EW-50126-24 Mfr # Y0-12NN
COMPARE
3/4 3/4 3/4
$38.80USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Nylon, 1" ID; 10/PK EW-50126-25 Mfr # Y0-16NN
COMPARE
1 1 1
$47.90USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Nylon, 1/16" ID; 10/PK EW-50126-26 Mfr # Y0-1NN
COMPARE
1/16 1/16 1/16
$12.70USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Nylon, 5/32" ID; 10/PK EW-50126-27 Mfr # Y0-2.5NN
COMPARE
5/32 5/32 5/32
$21.10USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Nylon, 1/8" ID; 10/PK EW-50126-28 Mfr # Y0-2NN
COMPARE
1/8 1/8 1/8
$12.70USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Nylon, 3/16" ID; 10/PK EW-50126-29 Mfr # Y0-3NN
COMPARE
3/16 3/16 3/16
$21.10USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Nylon, 1/4" ID; 10/PK EW-50126-30 Mfr # Y0-4NN
COMPARE
1/4 1/4 1/4
$22.80USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Nylon, 5/16" ID; 10/PK EW-50126-31 Mfr # Y0-5NN
COMPARE
5/16 5/16 5/16
$27.40USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Nylon, 3/8" ID; 10/PK EW-50126-32 Mfr # Y0-6NN
COMPARE
3/8 3/8 3/8
$27.40USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Nylon, 1/2" ID; 10/PK EW-50126-33 Mfr # Y0-8NN
COMPARE
1/2 1/2 1/2
$28.60USD / Pkg of 10
Showing 1- 12 of 12