Eldon James Hose Barb Fittings, Y Union, Stainless Steel
Excellent for high temperature and harsh physical environments
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Quick and easy assembly without tools
  • Single barb design tightly “grips” the tubing
  • RoHS compliant
Easily combine or split fluid flow using Y unions. These durable stainless steel fittings perform well in high temperatures and challenging environments. Made of 316L molybdenum bearing stainless steel, they are widely used in pharmaceutical, food, and beverage applications. A single barb on each end of the fitting creates a tight seal to keep the tubing in place.
$20.80 - $66.00USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Barb Size / Tubing Id (Port 2) (in) Connection Type
Availability Pricing
Eldon James Hose Barb Fitting, Y Union, Stainless Steel, 5/32" ID; 1/Each EW-50127-97 Mfr # Y0-2.5SS
COMPARE
5/32 5/32 Hose Barb Union
$20.80USD / Each
Eldon James Hose Barb Fitting, Y Union, Stainless Steel, 3/16" ID; 1/Each EW-50127-98 Mfr # Y0-3SS
COMPARE
3/16 3/16 Hose Barb Union
$30.20USD / Each
Eldon James Hose Barb Fitting, Y Union, Stainless Steel, 5/16" ID; 1/Each EW-50127-99 Mfr # Y0-5SS
COMPARE
5/16 5/16 Hose Barb Union
$57.60USD / Each
Eldon James Hose Barb Fitting, Y Union, Stainless Steel, 3/8" ID; 1/Each EW-50128-00 Mfr # Y0-6SS
COMPARE
3/8 3/8 Hose Barb Union
$62.00USD / Each
Eldon James Hose Barb Fitting, Y Union, Stainless Steel, 1/2" ID; 1/Each EW-50128-01 Mfr # Y0-8SS
COMPARE
1/2 1/2 Hose Barb Union
$66.00USD / Each
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 5/32" ID; 10/PK EW-50128-97 Mfr # Y0-2.5NK7
COMPARE
5/32 5/32 Hose Barb Y
$37.60USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 1/8" ID; 10/PK EW-50128-98 Mfr # Y0-2NK7
COMPARE
1/8 1/8 Hose Barb Y
$25.20USD / Pkg of 10
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 3/16" ID; 10/PK EW-50128-99 Mfr # Y0-3NK7
COMPARE
3/16 3/16 Hose Barb Y
$37.60USD / Pkg of 10
Showing 1- 8 of 8