Eldon James SeriesLock® Quick-Disconnect Fittings, Large Hose Barb & Sanitary Flange Body, Kynar® PVDF
Specially designed quick-disconnect fitting provides a high-volume flow at low line pressures
  • Spring-free flow path® design
  • Audible click ensures secure connection
  • Safety slide release minimizes accidental disconnects
  • Sterile quick disconnect
  • High flow yet compact to fit in tight spaces
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
The SeriesLock® quick disconnect fittings with spring-free fluid path allow a high-volume flow while minimizing turbulence, foaming, and bubbles. The SeriesLock® quick disconnect fittings with spring-free fluid path allow a high-volume flow while minimizing turbulence, foaming, and bubbles. For a secure, leak-free connection, push a large fitting insert into a large fitting body until you hear an audible “click.” Simply use the safety slide release to disconnect the fittings. The slide release eliminates accidental separation while offering easy disconnect with one hand even while wearing gloves. Recessed connection features protect contact surfaces and prolong the life of the O-rings.
Kynar PVDF quick-disconnect fittings are biocompatible and exhibit excellent UV, chemical, corrosion, and abrasion resistance. Transparent fittings allow easy viewing of fluid flow. Use fittings in temperatures ranging from 0 to 121°C (32 to 250°F).
$16.20 - $44.80USD / Each
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Connection Type
Availability Pricing
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Body, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 1/2" ID EW-50138-61 Mfr # B-FV-KO-VS-8
COMPARE
1/2 Quick-Disconnect Body x Hose Barb
$31.20USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Body, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 5/8" ID EW-50138-62 Mfr # B-FV-KO-VS-10
COMPARE
5/8 Quick-Disconnect Body x Hose Barb
$32.30USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Body, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 3/4" ID EW-50138-63 Mfr # B-FV-KO-VS-12
COMPARE
3/4 Quick-Disconnect Body x Hose Barb
$31.20USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Body, Kynar PVDF, Non-Valved, Viton O-Ring, SS Spring, Large, 1/2" ID EW-50138-74 Mfr # B-FO-KO-VS-8
COMPARE
1/2
$16.20USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Body, Kynar PVDF, Non-Valved, Viton O-Ring, SS Spring, Large, 5/8" ID EW-50138-75 Mfr # B-FO-KO-VS-10
COMPARE
5/8 Quick-Disconnect Body x Hose Barb
$16.20USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Hose Barb Body, Kynar PVDF, Non-Valved, Viton O-Ring, SS Spring, Large, 3/4" ID EW-50138-76 Mfr # B-FO-KO-VS-12
COMPARE
3/4 Quick-Disconnect Body x Hose Barb
$16.70USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 3/4" Sanitary & 1/2" ID EW-50139-24 Mfr # SF12-FV-KO-VS-8
COMPARE
1/2 Quick-Disconnect Body x 3/4" Sanitary Flange
$39.60USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 3/4" Sanitary & 5/8" ID EW-50139-25 Mfr # SF12-FV-KO-VS-10
COMPARE
5/8 Quick-Disconnect Body x 3/4" Sanitary Flange
$39.60USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 3/4" Sanitary & 3/4" ID EW-50139-26 Mfr # SF12-FV-KO-VS-12
COMPARE
3/4
$39.60USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Kynar PVDF, Non-Valved, Viton O-Ring, SS Spring, Large, 3/4" Sanitary & 1/2" ID EW-50139-37 Mfr # SF12-FO-KO-VS-8
COMPARE
1/2 Quick-Disconnect Body x 3/4" Sanitary Flange
$32.30USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Kynar PVDF, Non-Valved, Viton O-Ring, SS Spring, Large, 3/4" Sanitary & 5/8" ID EW-50139-38 Mfr # SF12-FO-KO-VS-10
COMPARE
5/8 Quick-Disconnect Body x 3/4" Sanitary Flange
$32.30USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Kynar PVDF, Non-Valved, Viton O-Ring, SS Spring, Large, 3/4" Sanitary & 3/4" ID EW-50139-39 Mfr # SF12-FO-KO-VS-12
COMPARE
3/4 Quick-Disconnect Body x 3/4" Sanitary Flange
$27.10USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 1-1/2" Sanitary & 1/2" ID EW-50139-60 Mfr # SF24-FV-KO-VS-8
COMPARE
1/2 Quick-Disconnect Body x 1-1/2" Sanitary Flange
$44.80USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 1-1/2" Sanitary & 5/8" ID EW-50139-61 Mfr # SF24-FV-KO-VS-10
COMPARE
5/8 Quick-Disconnect Body x 1-1/2" Sanitary Flange
$44.80USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Kynar PVDF, Valved, Viton O-Ring, SS Spring, Large, 1-1/2" Sanitary & 3/4" ID EW-50139-62 Mfr # SF24-FV-KO-VS-12
COMPARE
3/4 Quick-Disconnect Body x 1-1/2" Sanitary Flange
$44.80USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Kynar PVDF, Non-Valved, Viton O-Ring, SS Spring, Large, 1-1/2" Sanitary & 1/2" ID EW-50139-69 Mfr # SF24-FO-KO-VS-8
COMPARE
1/2 Quick-Disconnect Body x 1-1/2" Sanitary Flange
$33.30USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Kynar PVDF, Non-Valved, Viton O-Ring, SS Spring, Large, 1-1/2" Sanitary & 5/8" ID EW-50139-70 Mfr # SF24-FO-KO-VS-10
COMPARE
5/8
$33.30USD / Each
Eldon James SeriesLock® Quick-Disconnect Fitting, Sanitary Flange Body, Kynar PVDF, Non-Valved, Viton O-Ring, SS Spring, Large, 1-1/2" Sanitary & 3/4" ID EW-50139-71 Mfr # SF24-FO-KO-VS-12
COMPARE
3/4
$33.30USD / Each
Showing 1- 18 of 18