Encon Swing-Away Countertop Eyewash
Swing eyewash arm over sink to automatically activate spray
These easy-to-use Swing-Away eyewash arms can be mounted for either right- or left-handed use. Chemical-resistant acetal heads have flip-top covers; self-adjusting regulators provide a constant flow of 2.4 gpm at 3 psi. Supply connection is 1/2" NPT(M). Polished-chrome plating.
Compliance: ANSI Z358.1-2004
$713.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bowl Material Eyewash Fixture Material Mount
Availability Pricing
Encon 01040128 Swing-Away Countertop Eyewash, Arm Length; 10.5" EW-06767-82 Mfr # 01040128
COMPARE
Countertop Mount
$713.00USD / Each