Environmental Express® Chemically Preserved HDPE Bottles
EPA-approved containers with premeasured preservative already inside
  • Meets EPA specifications from "Guidance for Contaminant-Free Sample Containers" for metals, cyanide, fluoride, and other water quality parameters testing
  • Containers are lot controlled, sealed with custody tape, and include a Level 1 Certificate of Analysis from an independent laboratory
  • Included caps are composed of polypropylene (PP) and lined with F217, a low-density foam core between two solid layers of LDPE
$45.85 - $75.55USD / Case of 48
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Special Features
Availability Pricing
COMPARE
125 Filled with 0.5 mL of Sulfuric Acid
$45.85USD / Case of 48
COMPARE
250 Filled with 6 mL of Sodium Hydroxide
$47.15USD / Case of 24
COMPARE
500 Filled with 2 mL of Sodium Hydroxide
$75.55USD / Case of 24