Environmental Express® COD Standards
Controls approved for use in both EPA-compliant and mercury-free vials
  • Made from primary standard grade potassium hydrogen phthalate
  • Delivered with an SDS and minimum 8 month shelf life (refrigeration required)
  • Sold as one 250-mL bottle
  • Easy-to-follow dilution directions printed directly on the label
Primary Standard Grade Potassium Hydrogen Phthalate. Refrigeration is recommended.
$38.70USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
COMPARE
250 COD Standard, 1000 mg/L (ppm); 250 mL
$38.70USD / Each
COMPARE
250 COD Standard, 10000 mg/L (ppm); 250 mL
$38.70USD / Each