Environmental Express® HotBlock Hot Plate
Extra-large surface for heating multiple items at the same time
  • Uniform heating of the entire block
  • Simultaneously view both actual and set temperatures on the digital display
  • Type K thermocouple monitors heat settings
The corrosion-resistant HotBlock® hot plate was designed for environmental, as well as other, acid digestions prior to metals determinations. Compact size allows it to fit comfortably inside a fume hood. Large, low-wattage heating mat and provides extremely uniform temperatures to all parts of the block—great for glass and PTFE vessels. Durable graphite and Kydex® construction.
$4,501.90 - $4,590.70USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
COMPARE
120
$4,590.70USD / Each
COMPARE
240
$4,501.90USD / Each