Environmental Express® ICP Analytical Mixture Standards (8 Components), 100 µg/mL in 20% HCl
Multi-component analytical standards for ICP analysis
  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3100 series
  • For US EPA Methods 200.7 and 6010D
Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$242.00 - $271.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
100 µg/mL (100 ppm) 500 Analytical Mixture for ICP (8 Components), 100 µg/mL in 20% HCl; 500 mL
$271.00USD / Each
COMPARE
100 µg/mL (100 ppm) 250 Analytical Mixture for ICP (8 Components), 100 µg/mL in 20% HCl; 250 mL
$242.00USD / Each