Environmental Express® ICP Continuing Check Verification (CCV) Standards (25 Components) in 4% HNO3 + Tr HF, Solution A
Multi-component CCV standards for ICP analysis
  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3100 series
  • For US EPA Methods 200.7 and 6010D
Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$348.00 - $681.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
Multiple 500 Continuing Check Verification (CCV) Standard for ICP (25 Components) in 4% HNO3 & Tr HF, Solution A; 500 mL
$681.00USD / Each
COMPARE
Multiple 250 Continuing Check Verification (CCV) Standard for ICP (25 Components) in 4% HNO3 & Tr HF, Solution A; 250 mL
$483.00USD / Each
COMPARE
Multiple 100 Continuing Check Verification (CCV) Standard for ICP (25 Components) in 4% HNO3 & Tr HF, Solution A; 100 mL
$348.00USD / Each