Environmental Express® Magnesium, 10,000 µg/mL in 4% HNO3

Single-component standards for ICP analysis
  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3131
  • For AOAC Methods 990.08 and 975.03
Single-component standards offer high-quality, consistent results while saving preparation time.

Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.

Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.  
$104.00 - $267.00USD / Each

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
10,000 µg/mL(10,000 ppm) 100 Magnesium, 10,000 µg/mL for ICP in 4% HNO3; 100 mL
$104.00USD / Each
COMPARE
10,000 µg/mL(10,000 ppm) 250 Magnesium, 10,000 µg/mL for ICP in 4% HNO3; 250 mL
$173.00USD / Each
COMPARE
10,000 µg/mL(10,000 ppm) 500 Magnesium, 10,000 µg/mL for ICP in 4% HNO3; 500 mL
$267.00USD / Each
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat