Environmental Express® MicroBlock Distillation Systems

Perform up to ten cyanide, phenol, or ammonia distillations simultaneously
  • Reduce chemical waste by 90%
  • Get more accurate distillations--feature digital temperature and timer
  • Conform to EPA and Standard Methods
The Microblock systems all have an adjustable digital temperature control with LCD readout and timer. Includes a flow meter to monitor cooling water rates. The systems performs up to 10 distillations simultaneously.
$15,986.25 - $18,466.90USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
MicroBlock-V System with Cyanide Glassware; 120 V
$17,364.40USD / Each
COMPARE
MicroBlock-V System with Cyanide Glassware; 240V
$17,915.65USD / Each
COMPARE
MicroBlock-V System with Ammonia/Phenol Glassware; 120 V
$18,466.90USD / Each
COMPARE
MicroBlock-R System with Phenol/Ammonia Glassware; 120V
$15,986.25USD / Each
COMPARE
MicroBlock-R System with Phenol/Ammonia Glassware; 240V
$16,261.90USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat