Environmental Express® Oakton® 250 and 260 Series Handheld Meters
Reduce maintenance time with self-diagnostic function
  • Rugged, waterproof, and dustproof with an IP67 rating for use in any environment
  • Nonslip ergonomic design for comfortable handling
  • Easily view measurements with large monochrome LCD with white LED backlighting
  • Download and analyze data with USB output and phono jack printer connections
  • Take it with you—500 hours of battery life
Get easy-to-read and accurate readings with auto-hold, auto-stable, and real-time measurements with stability indicators. The Environmental Express® Oakton® 200 series of water quality meters features an electrode status indicator, which shows the condition of the electrode after each calibration. Easily view electrode condition, reading stability, measured parameter, and temperature in one screen. Don’t worry about accuracy in fluctuating temperature—ATC ensures measurement accuracy. The simple user interface features adjustable auto shut-off time ranging from 1 to 30 minutes. Self-diagnosing meters display measurement and meter issues for fast fault finding and troubleshooting.
Easily take measurements and conveniently store meter and electrodes with built-in foldable stand on the back of the meter. Two grooves on the side of the meter serve as electrode holders, to hold up to 2 electrodes.
Choose from two series—the 250 series feature 500 data points while the 260 series features 1000 data points, a real-time clock, and USB communication.
$508.00 - $1,720.00USD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Data Logging (points) Probe Included Description
Availability Pricing
COMPARE
500 Yes Waterproof Dual-Channel pH, ORP, Conductivity, TDS, Resistivity, and Salinity Handheld Meter Kit
$1,365.00USD / Each
COMPARE
1000 Yes Waterproof Dual-Channel pH, ORP, Conductivity, TDS, Resistivity, and Salinity Handheld Meter Kit
$1,410.00USD / Each
COMPARE
500 Yes Waterproof Dual-Channel pH, ORP, and DO Handheld Meter Kit
$1,565.00USD / Each
COMPARE
1000 Yes Waterproof Dual-Channel pH, ORP, and DO Handheld Meter Kit
$1,720.00USD / Each
COMPARE
500 No Waterproof pH and ORP Handheld Meter
$508.00USD / Each
COMPARE
1000 No Waterproof pH and ORP Handheld Meter
$665.00USD / Each
COMPARE
500 No Waterproof Conductivity, TDS, Resistivity, and Salinity Handheld Meter
$513.00USD / Each
COMPARE
1000 No Waterproof Conductivity, TDS, Resistivity, and Salinity Handheld Meter
$658.00USD / Each
COMPARE
500 No Waterproof Dual-Channel pH, ORP, Conductivity, TDS, Resistivity, and Salinity Handheld Meter
$711.00USD / Each
COMPARE
1000 No Waterproof Dual-Channel pH, ORP, Conductivity, TDS, Resistivity, and Salinity Handheld Meter
$868.00USD / Each
COMPARE
500 No Waterproof Dual-Channel pH, ORP, and DO Handheld Meter
$724.00USD / Each
COMPARE
1000 No Waterproof Dual-Channel pH, ORP, and DO Handheld Meter
$870.00USD / Each
Showing 1- 12 of 12