Environmental Express® Snip & Pour® Glucose-Glutamic Acid (GGA) Standard
Eliminate pipetting and dilutions
  • No more worries about sample contamination
  • Ready-to-use dilutions are easy and convenient
Each vial contains the APHA specified concentration of GGA. Simply shake the vial thoroughly, remove the seal and cap, pour contents, and rinse the vial at least two times with BOD water into the check standard solution. Product has a 24-month shelf life.
$42.30 - $80.00USD / Pkg of 24
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Buffer Kit Values Volume (mL) Description
Availability Pricing
COMPARE
24 vials 6 Snip & Pour® Glucose-Glutamic Acid (GGA), 24 x 6 mL Vials/PK
$80.00USD / Pkg of 24
COMPARE
24 ampules 1.2 Environmental Express Glucose-Glutamic Acid (GGA), 24 x 1.2 mL Ampules/Pk
$42.30USD / Pkg of 24