Environmental Express® Soil-Cell Systems
Replaces microwave technology for safer sample preparations
  • Complies with EPA Method 3546 soil extraction for organics
  • Ideal for PAH, BNA, TPH, and PCB compounds
The Soil-Cell® system simplifies the extraction of semi-volatile organic compounds in soil. The system meets the requirements to replace microwave technology with a HotBlock® unit and stainless steel extraction cells.
Place weighed sample into stainless steel extraction cell and add solvent. Tighten lid and heat at 130°C for 30 minutes. Allow cells to cool, then filter—extract is now ready for concentration, cleanup, and analysis.
$14,703.75 - $20,756.95USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number Of Blocks Power (VAC) Description
Availability Pricing
COMPARE
1 120 Soil-Cell System, 25-Well; 120 VAC
$14,703.75USD / Each
COMPARE
1 240 Soil-Cell System, 25-Well; 240 VAC
$15,735.65USD / Each
COMPARE
1 120 Soil-Cell System, 35-Well; 120 VAC
$20,756.95USD / Each
COMPARE
1 240 Soil-Cell System, 35-Well; 240 VAC
$20,736.90USD / Each