Environmental Express® TCLP Heavy-Duty LE Rotator
Greater degree of safety with increased reliability
  • Optimize your choice of extraction vessels
  • Requires no maintenance--self-lubricating, no-slip chain
  • Complies with EPA method 1311
Ideal for zero headspace and TCLP extractions, the durable twelve-position rotator is constructed with stainless steel, and has high-efficiency gear motor to ensure a long and reliable service life. Precise, constant, speed control is maintained during the long hours of operation.
Polyurethane extractor pads are available for a wide variety of vessels to fit the rotators. Styles are available for both zero headspace analyzers and TCLP bottles.
$8,625.75USD / Pkg of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Power (VAC)
Availability Pricing
COMPARE
30 120
$8,625.75USD / Pkg of 1
COMPARE
30 240
$8,625.75USD / Pkg of 1