Environmental Express® Ytterbium, 1,000 µg/mL in 2% HCl
Single-component standards for ICP analysis
  • ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited
  • Traceable to NIST SRM 3166
  • For ASTM Method D1976 - 20
Single-component standards offer high-quality, consistent results while saving preparation time.
Can’t find the standards you are looking for? HPS can make custom standards to meet your exact needs. Contact us for more information at [email protected] or call us at 843-767-7900.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$85.00 - $124.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Concentration Volume (mL) Description
Availability Pricing
COMPARE
1,000 µg/mL (1,000 ppm) 100 Ytterbium, 1,000 µg/mL for ICP in 2% HCl; 100 mL
$85.00USD / Each
COMPARE
1,000 µg/mL (1,000 ppm) 250 Ytterbium, 1,000 µg/mL for ICP in 2% HCl; 250 mL
$124.00USD / Each