Extech Digital Thermohygrometer with calibration
Optional thermistor probe lets you calculate T1 - T2
Compact meter is ideal for field applications — it displays relative humidity and temperature simultaneously, and dew point or wet bulb or probe temperature on a large LCD. Features include a choice of readings in degF or degC, Min/Max, data hold, and auto power off. Unique sensor cap closes to protect the sensor during storage. Advanced features include the calculation of T1-T2 temperature differential with external temperature probe 37803-18 (sold separately).
Unique sensor cap closes to protect the sensor during storage. Meter can be calibrated in the field using optional RH calibration standard salts. Order meter only or the complete thermohygrometer kit.
$114.00 - $184.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Battery Max RH Range (%) Max Temperature (° F)
Availability Pricing
Extech RH300 Digital Psychrometer, 0 to 100% RH/, -4 to 122F EW-37803-10 Mfr # RH300
COMPARE
2 AAA (included) 100 122
$114.00USD / Each
Extech RH350 Advance SuperHeat Digital Datalogging Psychrometer EW-37803-12 Mfr # RH350
COMPARE
4 AAA (included) 90 122
$184.00USD / Each