FLIR C-Series Pocket-Size Thermal Imager
Pocket portable—never miss an opportunity to uncover issues
  • Radiometric image capable of capturing thermal measurements from -4 to 752°F (-20 to 400°C)
  • MSX®-enhanced thermal images provide amazing detail to help you identify problem areas
  • Streaming video via FLIR Tools software
  • Bright 3.5” (9 cm) intuitive touch screen with auto orientation for easy viewing
The FLIR C-Series is a full-featured, pocket-sized thermal camera. Designed for contractors, building industry experts, and anyone that needs a pocket-sized thermal camera. The slim, light profile fits comfortably in your work pocket so you can always keep your camera at your side and never miss an opportunity to uncover issues. Use it as a nondestructive assessment tool and a reliable way to verify the success of completed repairs. The 19,200-pixel resolution, high-sensitivity detector captures and displays subtle thermal patterns and small temperature differences.
Full radiometric – the camera stores 5000 individual measurements on to each JPEG image that can be then be fully analyzed using FLIR Tools software.
Flir Tools software – with the included software you can analyze, change color palettes and access both thermal and visible imagery to create detailed reports.
MSX image enhancement – adds key details from the onboard visible light camera to the entire infrared image in real time. The result: an all-in-one, undiluted thermal picture with visible light features that lets you instantly recognize where the problematic heat pattern is.
$649.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Data Output Max Temperature (° F) Pixels / Thermal Resolution
Availability Pricing
Flir C5 Pocket Thermal Imaging Camera with MSX EW-39756-84 Mfr # FLIR C5
COMPARE
USB, Wi-Fi 752 19,200
$649.00USD / Each