FLIR® CM276 Thermal Imaging Clamp Meter Accessories
Enhance your CM276 thermal imaging clamp meter
  • Rated up to 1500 VAC and 1500 VDC
Use these test leads with CM276 thermal imaging clamp meter to measure the power output of solar panels (photovoltaic cells or PV).
$78.00 - $88.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
AC Max (Volts) DC Max (Volts) Description
Availability Pricing
FLIR® TA85 PV Test Leads for FLIR® CM276 Clamp Meter EW-39759-21 Mfr # TA85
COMPARE
1500 1500 PV Test Leads for FLIR® CM276 Clamp Meter
$88.00USD / Each
Flir® MC4 PV Plugs for FLIR® CM276 Clamp Meter EW-39759-22 Mfr # TA86
COMPARE
1500 1500 PV Plugs for FLIR® CM276 Clamp Meter
$78.00USD / Each