FLIR SV88™ and SV89™ Vibration Monitoring Accessories
FLIR’s accessory line for the SV88™ and SV89™ Vibration Monitoring Kits brings powerful scalability, remote access, and versatile mounting solutions
  • Stream real-time vibration data from multiple sensors through the powerful GW66 remote monitoring gateway
  • Swap out or expand coverage with additional SV88 or SV89 sensors for total equipment visibility
  • Mount sensors in seconds with the optional TA88 magnetic mount for fast, flexible deployment
  • Centralize and analyze all sensor data with seamless protocol support including Modbus, MQTT, and OPC UA
Take your vibration monitoring to the next level with FLIR’s high-performance accessories. The GW66 Gateway connects wirelessly to your SV88 and SV89 sensors, providing a unified platform for data aggregation, analysis, and real-time alerts—all from a secure, Linux-based web interface.
Need to cover more machines or replace a worn unit? Additional SV88 and SV89 sensors are built with the same rugged, IP66-rated design, and easily integrate into your existing system. And with the TA88 magnetic mount, installations become quicker and more adaptable—ideal for rotating equipment or areas with limited bolt access.
Whether you're expanding your system or optimizing what you already have, FLIR accessories give you the flexibility and reliability to maintain peak performance across your operation.
$67.00 - $589.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Flir SV88™ Vibration Monitoring and Temperature Sensor EW-39759-39 Mfr # SV88
COMPARE
SV88™ Vibration Monitoring and Temperature Sensor
$479.00USD / Each
Flir SV89™ Vibration Monitoring and Temperature Sensor EW-39759-40 Mfr # SV89
COMPARE
SV89™ Vibration Monitoring and Temperature Sensor
$589.00USD / Each
Flir Remote Monitoring Gateway for SV88 and SV89 EW-39759-41 Mfr # GW66
COMPARE
Remote Monitoring Gateway for SV88 and SV89
$539.00USD / Each
Flir Universal Power Adaptor for the GW66 Wireless Gateway EW-39759-42 Mfr # TA87
COMPARE
Universal Power Adaptor for the GW66 Wireless Gateway
$68.00USD / Each
Flir Magnetic Mount for SV88 and SV89 EW-39759-43 Mfr # TA88A
COMPARE
Magnetic Mount for SV88 and SV89
$67.00USD / Each