Fluke 59 Max and 59 Max+ Entry-Level Infrared Thermometers
Fits your job and your budget
  • Large, easy-to-read backlit LCD
  • Ergonomic design for a more natural hand fit
  • IP40 rated for extra protection
  • Durable—withstands a 3.3-ft (1-m) drop
Temperature is often the first indication of a potential problem. The Fluke 59 MAX noncontact thermometers are ideal for quick, basic temperature checks in applications where the user needs to be a safe distance from the target area being measured. These compact, portable tools enable professionals to scan surfaces for hot spots, diagnose problems, and monitor temperature variations without direct contact. Easy to use—just aim the laser sighting at the center of the target area, pull the trigger, and read the temperature on the large, backlit LCD.
Thermometers also feature selectable Fahrenheit or Celsius units, current/min/max temperature readings, average and differential temperatures, adjustable high/low set points with audible alarm, data hold, and power off.
ITEM(S) ON SALE
$115.20USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Distance To Target Size Ratio (D:S) Emissivity Max Temperature (° F)
Availability Pricing
Fluke 59 MAX Compact Infrared Thermometer (8: 1) EW-39641-07 Mfr # FLUKE-59 MAX NA
COMPARE
8:1 Adjustable 662
$115.20USD / Each
$128.00USD / Each