Fluke 725 and 726 Multi-Function Calibrators
  • Measure volts. mA, RTDs, thermocouples, frequency, and resistance to test sensors and transmitters
  • Source / simulate volts, mA, thermocouples, RTDs, frequency, and pressure to calibrate transmitters
  • Source mA with simultaneous pressure measurement to conduct valve and I/P tests
  • Perform fast linearity tests with auto step and auto ramp features
  • Power transmitters during test using 24 V loop supply and simultaneous mA measurement
  • Store frequently used test setups for later use
  • Available for sale in the United States and Canada only.
$5,611.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max Measurement (mA) Max Measurement (VDC)
Availability Pricing
Fluke 726 Precision Multi-Process Calibrator EW-16100-40 Mfr # FLUKE-726
COMPARE
$5,611.00USD / Each