Fluke® 750P Series Pressure Modules
Makes a multifunction or documenting calibrator into a pressure calibrator
  • High accuracy with reference uncertainty as good as 0.01%
  • Specifications at 6 months and 1 year with multiple temperature windows when better accuracies are needed
  • Sends digital communication to calibrators, eliminating analog losses and errors
  • 1/4" NPT, 1/4" BSP/ISO, and M20 male adapters are included for maximum compatibility
  • A rugged urethane overmolding protects and seals each module from shock, dirt, and moisture
  • Durable construction is backed by a 3-year warranty
These Fluke ® 750P series pressure modules turn your Fluke 753/754 Documenting Process Calibrators and 725/726 Multifunction Process Calibrators into pressure calibrators capable of operating with gauge, differential, dual range, absolute and vacuum pressures.
Each module incorporates a silicon piezo-resistive sensor and microprocessor that provides inherent temperature compensation and allows the module to communicate digitally to the calibrator. It also makes the modules fully interchangeable with a calibrator with each module having its own serial number and calibration traceability.
24 PRODUCT OPTIONS
Compareitem specifications
Max Pressure Min Pressure Pressure Accuracy
Availability Pricing
Fluke 750P00 Pressure Module, Differential, 0 to 1 Inch Water EW-16107-47 Mfr # FLUKE-750P00
COMPARE
1 inch of water (2.5 mbar) 0 inch of water (0 mbar) ±0.15%
$2,695.00USD / Each
Fluke 750P01 Pressure Module, Differential, 0 to 10 Inch Water EW-16107-48 Mfr # FLUKE-750P01
COMPARE
10 inch of water (25 mbar) 0 inch of water (0 mbar) ±0.1%
$2,338.00USD / Each
Fluke 750P23 Pressure Module, Differential, Stainless Steel, 0 to 5 psi EW-16107-50 Mfr # FLUKE-750P23
COMPARE
5 psi (350 mbar) 0 psi (0 mbar) ±0.02%
$2,338.00USD / Each
Fluke 750P04 Pressure Module, Differential, 0 to 15 psi EW-16107-51 Mfr # FLUKE-750P04
COMPARE
15 psi (1 bar) 0 psi (0 bar) ±0.0175%
$2,338.00USD / Each
Fluke 750P05 Pressure Module, Gage, 0 to 30 psi EW-16107-53 Mfr # FLUKE-750P05
COMPARE
30 psi (2.0 bar) 0 psi (0 bar) ±0.0175%
$2,338.00USD / Each
Fluke 750P06 Pressure Module, Gage, 0 to 100 psi EW-16107-54 Mfr # FLUKE-750P06
COMPARE
100 psi (7 bar) 0 psi (0 bar) ±0.0175%
$2,338.00USD / Each
Fluke 750P27 Pressure Module, Gage, 0 to 300 psi EW-16107-55 Mfr # FLUKE-750P27
COMPARE
300 psi (20 bar) 0 psi (0 bar) ±0.0175%
$2,338.00USD / Each
Fluke 750P07 Pressure Module, Gage, 0 to 500 psi EW-16107-56 Mfr # FLUKE-750P07
COMPARE
500 psi (35 bar) 0 psi (0 bar) ±0.0175%
$2,338.00USD / Each
Fluke 750P08 Pressure Module, Gage, 0 to 1000 psi EW-16107-57 Mfr # FLUKE-750P08
COMPARE
1000 psi (70 bar) 0 psi (0 bar) ±0.0175%
$2,338.00USD / Each
Fluke 750P2000 Pressure Module, Gage, 0 to 2000 psi EW-16107-59 Mfr # FLUKE-750P2000
COMPARE
2000 psi (140 bar) 0 psi (0 bar) ±0.0175%
$2,338.00USD / Each
Fluke 750P29 Pressure Module, Gage, 0 to 3000 psi EW-16107-60 Mfr # FLUKE-750P29
COMPARE
3000 psi (200 bar) 0 psi (0 bar) ±0.0175%
$3,403.00USD / Each
Fluke 750P30 Pressure Module, Gage, 0 to 5000 psi EW-16107-61 Mfr # FLUKE-750P30
COMPARE
5000 psi (340 bar) 0 psi (0 bar) ±0.0175%
$3,403.00USD / Each
Fluke 750P31 Pressure Module, Gage, 0 to 10,000 psi EW-16107-62 Mfr # FLUKE-750P31
COMPARE
10000 psi (700 bar) 0 psi (0 bar) ±0.0175%
$3,403.00USD / Each
Fluke 750PA4 Pressure Module, Absolute, 0 to 15 psi EW-16107-64 Mfr # FLUKE-750PA4
COMPARE
15 psia (1 bar) 0 psia (0 bar) ±0.03%
$2,612.00USD / Each
Fluke 750PA5 Pressure Module, Absolute, 0 to 30 psi EW-16107-65 Mfr # FLUKE-750PA5
COMPARE
30 psia (2.0 bar) 0 psia (0 bar) ±0.03%
$2,612.00USD / Each
Fluke 750PA6 Pressure Module, Absolute, 0 to 100 psi EW-16107-66 Mfr # FLUKE-750PA6
COMPARE
100 psia (7 bar) 0 psia (0 bar) ±0.03%
$2,612.00USD / Each
Fluke 750PD2 Pressure Module, Dual, -1 to 1 psi EW-16107-73 Mfr # FLUKE-750PD2
COMPARE
1 psi (70 mbar) ­1 psi (-70 mbar) ± 0.05%
$2,612.00USD / Each
Fluke 750PD3 Pressure Module, Dual, -5 to 5 psi EW-16107-74 Mfr # FLUKE-750PD3
COMPARE
5 psi (350 mbar) ­5 psi (-350 mbar) ±0.03%
$2,612.00USD / Each
Fluke 750PD4 Pressure Module, Dual, -15 to 15 psi EW-16107-76 Mfr # FLUKE-750PD4
COMPARE
15 psi (1 bar) ­15 psi (-1 bar) ±0.0175%
$2,612.00USD / Each
Fluke 750PD6 Pressure Module, Dual, -15 to 100 psi EW-16107-79 Mfr # FLUKE-750PD6
COMPARE
100 psi (7 bar) ­15 psi (-1 bar) ±0.0175%
$2,612.00USD / Each
Showing 1 - 20 of 24