Fluke Airflow Meter
The better way to measure air flow
  • Powerful meter provides differential and static pressure, air velocity and flow readings
  • Rugged design built for field use
  • Easy to use without sacrificing performance
  • User-defined duct shape and size for maximum airflow accuracy
  • Convenient colored hoses help you diagnose pressure readings
  • Bright, backlit display for clear viewing in all environments
  • Min/Max/Average/Hold functions for easy data analysis
  • Auto power-off saves battery life
Today’s HVAC technicians need a simple solution for diagnosing differential pressure and airflow ventilation issues. The Fluke 922 makes airflow measurements easy by combining pressure, air flow, and velocity into a single, rugged meter. Compatible with most pitot tubes, the Fluke 922 allows technicians to conveniently enter duct shape and dimensions for maximum measurement accuracy.
ITEM(S) ON SALE
$311.40 - $955.80USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Air Velocity Accuracy Max Air Velocity (ft/min) Description
Availability Pricing
Fluke 922 Airflow Meter / Digital Micromanometer EW-26048-99 Mfr # FLUKE-922
COMPARE
±2.5% of reading at 2000 ft/min (10.00 m/s) 16000 Airflow Meter / Digital Micromanometer
$955.80USD / Each
$1,062.00USD / Each
Fluke PT12 12 Inch Pitot Tube for 922 EW-26049-05 Mfr # PT12
COMPARE
12 Inch Pitot Tube for 922
$311.40USD / Each
$346.00USD / Each