Fluke Calibration Black Stack Readout Reference Thermometer
Offers accurate, expandable, and configurable readouts
  • Able to read SPRTs, RTDs, thermistors, and thermocouples
  • Can be configured with up to eight modules
  • Serves as a high-accuracy reference thermometer, with accuracy up to ±0.0013 °C
The 1560 Black Stack Thermometer Readout can be any kind of thermometer you want it to be, and it works in three distinctive ways. It’s a reference thermometer with a NIST traceable calibration; it’s an automated calibration system reading your reference probe and sensors you’re testing; or it’s a high-accuracy data acquisition system.
The system consists of up to eight different modules (sold separately) designed to do any type of thermometry you choose. Each module stacks behind the preceding one, and when you add a module, the Stack’s software automatically reconfigures itself to include all of the new functions supplied by that module. There’s no software to load, and nothing has to be calibrated. Each module stores its own calibration internally, so you can add or change modules without recalibrating the whole stack. Module calibration is digital and is performed manually through the base’s front panel or over the RS-232 link.
The LCD screen has multiple methods of displaying data, including a graphical strip chart recorder. The graphical capability of the Black Stack makes testing temperature stability easier than ever. Vertical scaling and graph resolution are automatic. Read data almost anyway you like—in ohms, millivolts, or temperature, according to your application and preference.
$1,905.00 - $4,651.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Fluke Calibration 2560 SPRT Module, 25 to 100 Ohm, 2-Channel EW-16101-40 Mfr # 2560
COMPARE
SPRT Module, 2-Channel
$4,651.00USD / Each
Fluke Calibration 9935-M Multi Channel LogWare Software EW-16103-16 Mfr # 9935-M
COMPARE
Software Channel Multi User
$1,905.00USD / Each